Assay Details
Target Gene Details
Entrez Gene ID: | 225845 |
Gene Name: | phospholipase A2, group XVI |
Gene Aliases: |
C78643, HRSL3, Hrasls3, Hrev107, MLP-3 |
Location: |
Chr.19:7557434-7588545 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pla2g16 | NM_139269.2 | 4 | 454 | NP_644675.2 |
| AB298806.1 | 2 | 299 | ||
| AK039703.1 | 4 | 667 | BAE20550.1 | |
| AK042007.1 | 4 | 407 | BAE20615.1 | |
| AK052657.1 | 4 | 433 | BAC35085.1 | |
| AK163505.1 | 4 | 455 | BAE37375.1 | |
| AX775891.1 | 4 | 436 | CAE11645.1 | |
| BC024581.1 | 4 | 400 | AAH24581.1 |

Genomic Map