Assay Details
Target Gene Details
Entrez Gene ID: | 225845 |
Gene Name: | phospholipase A2, group XVI |
Gene Aliases: |
C78643, HRSL3, Hrasls3, Hrev107, MLP-3 |
Location: |
Chr.19:7557434-7588545 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pla2g16 | NM_139269.2 | 5 | 1527 | NP_644675.2 |
| AK039703.1 | 5 | 1740 | BAE20550.1 | |
| AK042007.1 | 5 | 1480 | BAE20615.1 | |
| AK163505.1 | 5 | 1528 | BAE37375.1 |

Genomic Map