Assay Details
Target Gene Details
Entrez Gene ID: | 245386 |
Gene Name: | transmembrane protein 255A |
Gene Aliases: |
4933417N17, 6430550H21Rik, Fam70a |
Location: |
Chr.X:38196573-38252489 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem255a | NM_001289727.1 | NP_001276656.1 | ||
| NM_001289728.1 | 9 | 2570 | NP_001276657.1 | |
| NM_172930.4 | 10 | 2639 | NP_766518.3 | |
| AK041985.1 | BAC31122.1 | |||
| AK045375.1 | 8 | 2913 | BAC32335.1 | |
| AK078264.1 | 9 | 2515 | BAC37196.1 | |
| BC062956.1 | 10 | 2555 | AAH62956.1 |

Genomic Map