Assay Details
Target Gene Details
Entrez Gene ID: | 70999 |
Gene Name: | N(alpha)-acetyltransferase 40, NatD catalytic subunit, homolog (S. cerevisiae) |
Gene Aliases: |
4931433E08Rik, AU023197, Nat11 |
Location: |
Chr.19:7225668-7241222 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Naa40 | NM_027643.1 | 8 | 2257 | NP_081919.1 |
| AK033510.1 | 8 | 2079 | ||
| AK033896.1 | 8 | 2088 | BAC28506.1 | |
| AK035418.1 | 7 | 2975 | BAE20496.1 | |
| AK162625.1 | 7 | 2351 | ||
| AK170548.1 | 8 | 2083 | BAE41873.1 | |
| BC020020.1 | 8 | 2108 | AAH20020.1 | |
| BC058212.1 | 3 | 1916 | AAH58212.1 |

Genomic Map