Assay Details
Target Gene Details
Entrez Gene ID: | 56626 |
Gene Name: | polymerase (DNA directed), lambda |
Gene Aliases: |
1110003P06Rik, AV007317 |
Location: |
Chr.19:45552275-45560543 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Poll | NM_020032.2 | 3 | 292 | NP_064416.1 |
| AF176099.1 | 2 | 233 | AAF27553.1 | |
| AJ131889.1 | 3 | 292 | CAB65241.1 | |
| AK133087.1 | 3 | 259 | ||
| BC004767.1 | 2 | 242 | AAH04767.1 |
More Information
Set Membership: |
|

Genomic Map