Assay Details
Target Gene Details
Entrez Gene ID: | 18241 |
Gene Name: | G protein-coupled receptor 143 |
Gene Aliases: |
Oa1 |
Location: |
Chr.X:152781921-152808651 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpr143 | NM_010951.3 | 1 | 250 | NP_035081.3 |
| AK033060.1 | 1 | 154 | BAC28141.1 | |
| BC119384.1 | 1 | 166 | ||
| BC137813.1 | 1 | 166 | ||
| CJ131873.1 | 1 | 251 | ||
| U63918.1 | 1 | 185 | AAC52890.1 |
More Information
Set Membership: |
|

Genomic Map