Assay Details
Target Gene Details
Entrez Gene ID: | 67062 |
Gene Name: | solute carrier family 25, member 53 |
Gene Aliases: |
2310046F18Rik, 2810402A17Rik, AI448995, AI841128, Mcart6 |
Location: |
Chr.X:136981116-137038302 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc25a53 | NM_001082412.2 | NP_001075881.1 | ||
| AK076279.1 |
Target Gene Details
Entrez Gene ID: | 66995 |
Gene Name: | zinc finger, CCHC domain containing 18 |
Gene Aliases: |
1500031H04Rik, Sizn2 |
Location: |
Chr.X:136992781-136996923 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zcchc18 | NM_001035509.1 | 4 | 2444 | NP_001030586.1 |
| NM_001035510.1 | 2 | 2915 | NP_001030587.1 | |
| NM_025893.2 | 4 | 2392 | NP_080169.2 | |
| AV174394.2 | 1 | 189 |
More Information
Set Membership: |
|

Genomic Map