Assay Details
Target Gene Details
Entrez Gene ID: | 20592 |
Gene Name: | lysine (K)-specific demethylase 5D |
Gene Aliases: |
HY, Jarid1d, Smcy |
Location: |
Chr.Y:897660-943813 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kdm5d | NM_011419.3 | 23 | 4102 | NP_035549.1 |
| AB221606.1 | 9 | 1755 | ||
| AF127244.1 | 23 | 3940 | AAD53048.1 | |
| BC059077.1 | 15 | 2917 | AAH59077.1 | |
| BQ921842.1 | 1 | 84 |
More Information
Set Membership: |
|

Genomic Map