Assay Details
Target Gene Details
Entrez Gene ID: | 24071 |
Gene Name: | synaptojanin 2 binding protein |
Gene Aliases: |
AA409442, ARIP2, ARIP2a, ActRIP4, D12Wsu118e, OMP25 |
Location: |
Chr.12:81492194-81532907 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Synj2bp | NM_025292.6 | 4 | 5901 | NP_079568.1 |
More Information
Set Membership: |
|

Genomic Map