Assay Details
Target Gene Details
Entrez Gene ID: | 20723 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade B, member 9 |
Gene Aliases: |
BB283241, CAP-3, CAP3, PI-9, PI9, Spi6, ovalbumin |
Location: |
Chr.13:32997061-33017955 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpinb9 | NM_009256.3 | 7 | 1189 | NP_033282.1 |
| AK035624.1 | 7 | 1128 | BAC29128.1 | |
| AK087785.1 | 7 | 1099 | ||
| AK161581.1 | 7 | 1190 | BAE36475.1 | |
| BC029900.1 | 7 | 1117 | AAH29900.1 | |
| U96700.1 | 7 | 1068 | AAB57812.1 |
More Information
Set Membership: |
|

Genomic Map