Assay Details
Target Gene Details
Entrez Gene ID: | 72567 |
Gene Name: | BCL2-associated transcription factor 1 |
Gene Aliases: |
2610102K23Rik, 2700025J07Rik, 2810454G14Rik, 5730534O06Rik, AI450190, AW556225, Btf, mKIAA0164 |
Location: |
Chr.10:20311461-20342503 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bclaf1 | NM_001025392.1 | 13 | 3341 | NP_001020563.1 |
| NM_001025393.1 | 12 | 3194 | NP_001020564.1 | |
| NM_153787.2 | 13 | 3335 | NP_722482.1 | |
| AK013343.1 | 1 | 164 | ||
| AK079110.1 | 9 | 1591 | BAC37542.1 | |
| AK129071.1 | 10 | 2625 | BAC97881.1 | |
| AK169491.1 | 2 | 490 | BAE41198.1 | |
| CN530237.1 |
More Information
Set Membership: |
|

Genomic Map