Assay Details
Target Gene Details
Entrez Gene ID: | 238406 |
Gene Name: | a disintegrin and metallopeptidase domain 6A |
Gene Aliases: |
Adam6, tMDCIV |
Location: |
Chr.12:113543908-113546414 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adam6a | NM_174885.3 | 1 | 941 | NP_777479.2 |
| AY158689.1 | 1 | 941 | AAN77878.1 | |
| BC108387.1 | 1 | 894 | AAI08388.1 | |
| BC139059.1 | 1 | 926 |
More Information
Set Membership: |
|

Genomic Map