Assay Details
Target Gene Details
Entrez Gene ID: | 18750 |
Gene Name: | protein kinase C, alpha |
Gene Aliases: |
AI875142, Pkca |
Location: |
Chr.11:107933387-108343888 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prkca | NM_011101.3 | 17 | 8152 | NP_035231.2 |
| AK079091.1 | 1 | 3347 | ||
| BF455952.1 | 1 | 234 |
More Information
Set Membership: |
|

Genomic Map