Assay Details
Target Gene Details
Entrez Gene ID: | 214321 |
Gene Name: | predicted gene 4787 |
Gene Aliases: |
Adam4b, EG214321 |
Location: |
Chr.12:81376991-81379486 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm4787 | NM_001038995.2 | 1 | 1360 | NP_001034084.2 |
| BC141094.1 | 1 | 1338 | ||
| KF144384.1 | ||||
| KF144385.1 |
More Information
Set Membership: |
|

Genomic Map