Assay Details
Target Gene Details
Entrez Gene ID: | 11498 |
Gene Name: | a disintegrin and metallopeptidase domain 4 |
Gene Aliases: |
AV274654, tMDCV |
Location: |
Chr.12:81419549-81421878 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adam4 | NM_009620.1 | 1 | 872 | NP_033750.1 |
| AK076647.1 | 1 | 932 | ||
| AY158688.1 | 1 | 872 | AAN77877.1 | |
| BC150159.1 | 1 | 876 | ||
| BC150481.1 | 1 | 876 | ||
| U22058.1 | AAA74922.1 |

Genomic Map