Assay Details
Target Gene Details
Entrez Gene ID: | 66589 |
Gene Name: | ubiquitin-conjugating enzyme E2 variant 1 |
Gene Aliases: |
0610011J09Rik, AI256840, CROC-1, CROC1, D7Bwg1382e, UEV-1 |
Location: |
Chr.2:167607639-167632047 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ube2v1 | NM_023230.2 | 4 | 1812 | NP_075719.1 |
| AK158230.1 | 4 | 1812 | BAE34414.1 |
More Information
Set Membership: |
|

Genomic Map