Assay Details
Target Gene Details
Entrez Gene ID: | 229672 |
Gene Name: | BCLl2-like 15 |
Gene Aliases: |
Bfk, Gm566 |
Location: |
Chr.3:103831039-103854618 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bcl2l15 | NM_001142960.1 | 4 | 1841 | NP_001136432.1 |
| AV381247.2 | 1 | 507 |

Genomic Map