Assay Details
Target Gene Details
Entrez Gene ID: | 22190 |
Gene Name: | ubiquitin C |
Gene Aliases: |
2700054O04Rik, AI194771, Rps27a, TI-225, Uba52, Ubb |
Location: |
Chr.5:125385965-125390017 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ubc | NM_019639.4 | 2 | 2268 | NP_062613.3 |
| AI839133.1 | 1 | 157 | ||
| AK004727.1 | 3 | 505 | ||
| AK010964.1 | 4 | 1079 | BAB27296.2 | |
| AK168448.1 | 3 | 851 | BAE40351.1 | |
| BC006680.1 | 4 | 1750 | AAH06680.1 | |
| BC008661.1 | 3 | 633 | AAH08661.1 | |
| BC021837.1 | 3 | 1977 | AAH21837.1 | |
| BC025894.1 | 3 | 1070 | AAH25894.1 | |
| BC036303.1 | 3 | 1050 | AAH36303.1 | |
| BC094012.1 | 3 | 1066 | AAH94012.1 | |
| CO044895.1 | 1 | 147 | ||
| D50527.1 | 3 | 370 | BAA09096.1 | |
| S40697.1 | 1 | 99 | AAB22649.1 |

Genomic Map