Assay Details
Target Gene Details
Entrez Gene ID: | 66511 |
Gene Name: | chromatin target of PRMT1 |
Gene Aliases: |
2500003M10Rik, Fop, Srag |
Location: |
Chr.3:90498539-90509888 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chtop | NM_001293776.1 | 6 | 1920 | NP_001280705.1 |
| NM_001293777.1 | 6 | 1917 | NP_001280706.1 | |
| NM_001293778.1 | 5 | 1782 | NP_001280707.1 | |
| NM_001293779.1 | 5 | 1779 | NP_001280708.1 | |
| NM_001293780.1 | 4 | 3065 | NP_001280709.1 | |
| NM_001293781.1 | 4 | 3062 | NP_001280710.1 | |
| NM_001293782.1 | 6 | 2917 | NP_001280711.1 | |
| NM_023215.6 | 5 | 3203 | NP_075704.2 | |
| AB041654.1 | 4 | 3017 | BAA95102.1 | |
| AK004558.2 | 5 | 3149 | BAB23371.1 | |
| AK005377.1 | 5 | 1744 | BAB23984.2 | |
| AK008885.1 | 3 | 1216 | BAB25951.1 | |
| AK047914.1 | 6 | 1894 | BAC33189.1 | |
| AK132769.1 | 6 | 1893 | BAE21348.1 | |
| AK149010.1 | 3 | 1391 | BAE28718.1 | |
| BC004582.1 | 1 | 1052 | AAH04582.1 | |
| BC083333.1 | 4 | 1506 | AAH83333.1 | |
| BC108396.1 | 5 | 1549 | AAI08397.1 | |
| BE852996.1 |
More Information
Set Membership: |
|

Genomic Map