Assay Details
Target Gene Details
Entrez Gene ID: | 51813 |
Gene Name: | cyclin C |
Gene Aliases: |
AI451004, AU020987, CG1C |
Location: |
Chr.4:21727701-21759922 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccnc | NM_001290420.1 | NP_001277349.1 | ||
| NM_001290422.1 | 13 | 1139 | NP_001277351.1 | |
| NM_016746.4 | 12 | 1045 | NP_058026.2 | |
| AK014079.1 | 11 | 894 | ||
| AK037957.1 | 13 | 1140 | BAC29908.1 | |
| AK135472.1 | BAE22544.1 | |||
| BC003344.1 | 12 | 978 | ||
| BC100396.1 | 12 | 1019 | AAI00397.1 | |
| BC120649.1 | 12 | 972 | AAI20650.1 | |
| BC120677.1 | 12 | 972 | AAI20678.1 | |
| U62638.1 | 12 | 1020 | AAB05260.1 |
More Information
Set Membership: |
|

Genomic Map