Assay Details
Target Gene Details
Entrez Gene ID: | 26570 |
Gene Name: | solute carrier family 7 (cationic amino acid transporter, y+ system), member 11 |
Gene Aliases: |
9930009M05Rik, AI451155, sut, xCT |
Location: |
Chr.3:50364936-50499087 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc7a11 | NM_011990.2 | 12 | 8929 | NP_036120.1 |
| AK051605.1 | 1 | 2802 | ||
| AY766236.1 | 12 | 8896 | AAX10866.1 |
More Information
Set Membership: |
|

Genomic Map