Assay Details
Target Gene Details
Entrez Gene ID: | 70599 |
Gene Name: | sperm specific antigen 2 |
Gene Aliases: |
5730488C15Rik, CS-1, CS1, KRAP, SPAG13, mKIAA1927 |
Location: |
Chr.2:79635373-79672966 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ssfa2 | NM_080558.4 | 11 | 2790 | NP_542125.3 |
| AB093303.1 | 11 | 2665 | BAC41485.1 | |
| AB120565.1 | 11 | 2675 | BAC87833.1 | |
| AK037213.1 | 9 | 2500 | BAC29756.1 | |
| AK169543.1 | 11 | 2791 | BAE41220.1 | |
| BC008560.1 | 4 | 1763 | AAH08560.1 | |
| BC018526.1 | ||||
| BC079535.1 | 5 | 2169 | AAH79535.1 | |
| BC115799.1 | ||||
| BC122519.1 | 11 | 2676 | ||
| BC141883.1 | 11 | 2675 |

Genomic Map