Assay Details
Target Gene Details
Entrez Gene ID: | 69726 |
Gene Name: | SET and MYND domain containing 3 |
Gene Aliases: |
2410008A19Rik, Zmynd1 |
Location: |
Chr.1:178951960-179518052 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smyd3 | NM_027188.3 | 12 | 1470 | NP_081464.1 |
| AK010447.1 | 12 | 1504 | BAB26947.1 | |
| AK043988.1 | 12 | 1480 | ||
| AK044168.1 | 8 | 1031 | BAC31804.1 | |
| AK046829.1 | 9 | 1077 | BAC32887.1 | |
| AK163057.1 | 12 | 1487 | BAE37175.1 | |
| AK167875.1 | 12 | 1485 | BAE39889.1 | |
| BC052431.1 | 12 | 1470 | AAH52431.1 | |
| BC061485.1 | 12 | 1453 | AAH61485.1 |
More Information
Set Membership: |
|

Genomic Map