Assay Details
Target Gene Details
Entrez Gene ID: | 230125 |
Gene Name: | solute carrier family 25, member 51 |
Gene Aliases: |
9130208E07Rik, D130005A03Rik, Gm138, Mcart1 |
Location: |
Chr.4:45395923-45408783 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc25a51 | NM_001009949.3 | 3 | 2172 | NP_001009949.3 |
| AK146788.1 | 3 | 2134 | BAE27432.1 | |
| AK156168.1 | 3 | 2130 | BAE33610.1 | |
| AK156342.1 | 3 | 2130 | ||
| AK164726.1 | 3 | 2153 | ||
| BC089000.1 | 2 | 2067 | AAH89000.1 |

Genomic Map