Assay Details
Target Gene Details
Entrez Gene ID: | 110805 |
Gene Name: | forkhead box E1 |
Gene Aliases: |
Titf2 |
Location: |
Chr.4:46344194-46345309 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Foxe1 | NM_183298.1 | 1 | 981 | NP_899121.1 |
More Information
Set Membership: |
|

Genomic Map