Assay Details
Target Gene Details
Entrez Gene ID: | 13205 |
Gene Name: | DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 3, X-linked |
Gene Aliases: |
D1Pas1-rs2, Ddx3, Fin14 |
Location: |
Chr.X:13280496-13293983 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddx3x | NM_010028.3 | 17 | 3018 | NP_034158.1 |
| AK140990.1 | 17 | 3019 | BAE24540.1 | |
| AK163250.1 | BAE37257.1 | |||
| AK211517.1 | 1 | 232 | ||
| BC150862.1 | ||||
| BC172016.1 | ||||
| DQ858457.1 | 7 | 1807 | ||
| DQ858458.1 | 4 | 1338 | ||
| DQ858459.1 | 3 | 1132 | ||
| U42386.1 | 1 | 621 | AAB08868.1 | |
| Z38117.1 | 17 | 3002 | CAA86261.1 |

Genomic Map