Assay Details
Target Gene Details
Entrez Gene ID: | 14294 |
Gene Name: | formyl peptide receptor 3 |
Gene Aliases: |
ALX, Fpr-rs1, Fpr-s1, Fprl1, LXA4-R, Lxa4r |
Location: |
Chr.17:17970458-17971677 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fpr3 | NM_008042.2 | 1 | 1061 | NP_032068.2 |
More Information
Set Membership: |
|

Genomic Map