Assay Details
Target Gene Details
Entrez Gene ID: | 70620 |
Gene Name: | ubiquitin-conjugating enzyme E2 variant 2 |
Gene Aliases: |
1110021H13Rik, 4632410D19Rik, 5730524P06Rik, 6820402M05, AI848315, C81524, MMS2 |
Location: |
Chr.16:15550986-15594518 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ube2v2 | NM_001159351.1 | 3 | 5222 | NP_001152823.1 |
| NM_023585.4 | 4 | 5348 | NP_076074.2 | |
| AK003898.1 | 1 | 1382 |
More Information
Set Membership: |
|

Genomic Map