Assay Details
Target Gene Details
Entrez Gene ID: | 67768 |
Gene Name: | N-6 adenine-specific DNA methyltransferase 1 (putative) |
Gene Aliases: |
5830445C04Rik, Hemk2, Pred28 |
Location: |
Chr.16:87354185-87368649 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| N6amt1 | NM_001159331.1 | 5 | 560 | NP_001152803.1 |
| NM_026366.2 | 6 | 702 | NP_080642.1 | |
| AK012019.1 | 5 | 1826 | ||
| AK013667.1 | 2 | 134 | BAB28948.2 | |
| AK145617.1 | 6 | 654 | BAE26543.1 | |
| AK160298.1 | 6 | 665 | ||
| AY456393.1 | 6 | 702 | AAR19227.1 | |
| AY583759.1 | 5 | 604 | AAS94315.1 | |
| BC116393.1 | 6 | 672 | ||
| BC116394.1 | 6 | 672 |
More Information
Set Membership: |
|

Genomic Map