Assay Details
Target Gene Details
Entrez Gene ID: | 382007 |
Gene Name: | a disintegrin and metallopeptidase domain 26B |
Gene Aliases: |
- |
Location: |
Chr.8:43519864-43528137 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adam26b | NM_001009547.2 | 2 | 2306 | NP_001009547.1 |
More Information
Set Membership: |
|

Genomic Map