Assay Details
Target Gene Details
Entrez Gene ID: | 73936 |
Gene Name: | coiled-coil domain containing 175 |
Gene Aliases: |
4930403N07Rik |
Location: |
Chr.12:72100973-72185029 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc175 | NM_028687.1 | NP_082963.1 |
Target Gene Details
Entrez Gene ID: | 104771 |
Gene Name: | JNK1/MAPK8-associated membrane protein |
Gene Aliases: |
1200003C05Rik, 2310047L11Rik, Jamp |
Location: |
Chr.12:72085839-72101527 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Jkamp | NM_001205067.1 | 7 | 1708 | NP_001191996.1 |
| NM_024205.2 | 7 | 1726 | NP_077167.1 | |
| AK004555.1 | 7 | 1663 | BAC25088.1 | |
| AK013502.1 | 2 | 755 | BAB28884.1 |
More Information
Set Membership: |
|

Genomic Map