Assay Details
Target Gene Details
Entrez Gene ID: | 27364 |
Gene Name: | serine racemase |
Gene Aliases: |
Srs |
Location: |
Chr.11:74905047-74925798 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srr | NM_001163311.1 | NP_001156783.1 | ||
| NM_013761.4 | NP_038789.1 | |||
| AF148321.1 | AAF08701.1 | |||
| AK017235.1 | ||||
| AK028034.1 | BAC25712.1 | |||
| AK031687.1 | BAC27514.1 | |||
| AK043738.1 | BAC31637.1 | |||
| AK079343.1 | ||||
| AK157122.1 | BAE33968.1 | |||
| AK170096.1 | BAE41561.1 | |||
| AK170471.1 | ||||
| BC011164.1 | AAH11164.1 |
More Information
Set Membership: |
|

Genomic Map