Assay Details
Target Gene Details
Entrez Gene ID: | 53605 |
Gene Name: | nucleosome assembly protein 1-like 1 |
Gene Aliases: |
AA407126, AI256722, D10Ertd68e, NAP-1 |
Location: |
Chr.10:111473192-111498150 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nap1l1 | NM_015781.4 | 1 | 48 | NP_056596.1 |
| AK004633.1 | ||||
| AK007322.1 | BAE43200.1 | |||
| AK032896.1 | BAC28075.1 | |||
| AK136161.1 | BAE22850.1 | |||
| AK145766.1 | BAE26637.1 | |||
| AK152011.1 | ||||
| AK153531.1 | ||||
| AK169301.1 | BAE41057.1 | |||
| D12618.1 | BAA02142.1 |
More Information
Set Membership: |
|

Genomic Map