Assay Details
Target Gene Details
Entrez Gene ID: | 217480 |
Gene Name: | diacylglycerol kinase, beta |
Gene Aliases: |
6430574F24, 90kda, C630029D13Rik, DAGK2, DGK, DGK-beta, mKIAA0718 |
Location: |
Chr.12:37880168-38634239 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dgkb | NM_178681.4 | 2 | 405 | NP_848796.2 |
| AK032509.1 | 2 | 564 | ||
| AK044516.1 | 2 | 215 | BAC31960.1 | |
| AK047682.1 | 2 | 320 | BAC33123.2 | |
| AK083222.1 | 2 | 395 | BAC38814.1 | |
| AK139408.1 | 2 | 406 | BAE23997.1 | |
| AK161933.1 | 2 | 401 | BAE36642.1 | |
| BC070461.1 | 2 | 237 | AAH70461.1 |
More Information
Set Membership: |
|

Genomic Map