Assay Details
Target Gene Details
Entrez Gene ID: | 19146 |
Gene Name: | transmembrane protease, serine 15 |
Gene Aliases: |
Entk, Prss7 |
Location: |
Chr.16:78953008-79091097 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 25 - Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmprss15 | NM_008941.3 | NP_032967.1 | ||
| NM_178855.4 | NP_849186.2 | |||
| AK033582.1 | BAC28373.1 | |||
| AK038356.1 | BAC29973.1 | |||
| BC117917.1 | ||||
| BC117918.1 | ||||
| U73378.1 | AAB37317.1 |
More Information
Set Membership: |
|

Genomic Map