Assay Details
Target Gene Details
Entrez Gene ID: | 223970 |
Gene Name: | RMI2, RecQ mediated genome instability 2, homolog (S. cerevisiae) |
Gene Aliases: |
A630055G03Rik, Gm537, Gm929 |
Location: |
Chr.16:10835059-10843235 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rmi2 | NM_001162932.1 | 1 | 139 | NP_001156404.1 |
| AK143588.1 | 1 | 140 | BAE25452.1 |

Genomic Map