Assay Details
Target Gene Details
Entrez Gene ID: | 18203 |
Gene Name: | N-terminal Asn amidase |
Gene Aliases: |
- |
Location: |
Chr.16:13818834-13835450 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ntan1 | NM_010946.3 | 10 | 919 | NP_035076.1 |
| AA161752.1 | ||||
| AK041112.1 | BAC30825.1 | |||
| AK206842.1 | 1 | 77 | ||
| AK208313.1 | 1 | 77 | ||
| BC030172.1 | 10 | 917 | AAH30172.1 | |
| U57692.1 | 10 | 873 | AAB66490.1 |
Target Gene Details
Entrez Gene ID: | 94184 |
Gene Name: | pyridoxal-dependent decarboxylase domain containing 1 |
Gene Aliases: |
2210010A19Rik, AA415817, Kiaa0251-hp |
Location: |
Chr.16:13833161-13903145 on Build GRCm38 |
Assay Gene Location: | Within Intron 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdxdc1 | NM_001039533.2 | NP_001034622.1 | ||
| NM_001291017.1 | NP_001277946.1 | |||
| AK004611.1 | BAB23408.1 | |||
| AK054329.1 | BAC35732.1 | |||
| AK163696.1 | BAE37462.1 |
More Information
Set Membership: |
|

Genomic Map