Assay Details
Target Gene Details
Entrez Gene ID: | 320873 |
Gene Name: | cadherin 10 |
Gene Aliases: |
A830016G23Rik, C030003B10Rik, C030011H18Rik |
Location: |
Chr.15:18818888-19016206 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdh10 | NM_009865.2 | 11 | 2230 | NP_033995.1 |
| AF183946.1 | 11 | 1999 | AAL67951.1 | |
| AK138606.1 | 11 | 2232 | BAE23714.1 | |
| BC052056.1 | 11 | 1938 | AAH52056.1 | |
| BC055808.1 | 2 | 801 | AAH55808.1 | |
| BC062962.1 | 11 | 1938 | AAH62962.1 | |
| U69137.1 | 2 | 197 | AAB87708.1 |
More Information
Set Membership: |
|

Genomic Map