Assay Details
Target Gene Details
Entrez Gene ID: | 100039139 |
Gene Name: | coiled-coil domain containing 152 |
Gene Aliases: |
ENSMUSG00000071814 |
Location: |
Chr.15:3280099-3304743 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc152 | NM_001166063.2 | NP_001159535.1 |

Genomic Map