Assay Details
Target Gene Details
Entrez Gene ID: | 56274 |
Gene Name: | serine/threonine kinase 3 |
Gene Aliases: |
0610042I06Rik, MST, Mst2, Mst3, Ste20, mess1 |
Location: |
Chr.15:34875496-35155806 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stk3 | NM_019635.2 | NP_062609.2 | ||
| AF271361.1 | AAF75790.1 | |||
| AY058922.1 | AAL29682.1 | |||
| BC037440.1 | AAH37440.1 | |||
| BC049123.1 | AAH49123.2 | |||
| U28726.1 | AAA75300.1 |
More Information
Set Membership: |
|

Genomic Map