Assay Details
Target Gene Details
Entrez Gene ID: | 100042814 |
Gene Name: | apolipoprotein L 7d |
Gene Aliases: |
ENSMUSG00000073652, Gm15456, OTTMUSG00000022136 |
Location: |
Chr.1:71653335-71662843 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apol7d | NR_040308.1 | |||
| AK049041.1 | ||||
| AK052223.1 | 2 | 120 | ||
| AK135370.1 | BAE22508.1 |
More Information
Set Membership: |
|

Genomic Map