Assay Details
Target Gene Details
Entrez Gene ID: | 17936 |
Gene Name: | Ngfi-A binding protein 1 |
Gene Aliases: |
- |
Location: |
Chr.1:52455849-52500448 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nab1 | NM_008667.3 | NP_032693.2 | ||
| AB221677.1 | ||||
| BC005627.1 | AAH05627.1 | |||
| BC016886.1 | AAH16886.1 | |||
| U47008.1 | AAC52649.1 |
More Information
Set Membership: |
|

Genomic Map