Assay Details
Target Gene Details
Entrez Gene ID: | 384557 |
Gene Name: | carcinoembryonic antigen-related cell adhesion molecule 3 |
Gene Aliases: |
EG384557, Psg24, cea12 |
Location: |
Chr.7:17150195-17163754 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ceacam3 | NM_054059.1 | 3 | 468 | NP_473400.1 |
More Information
Set Membership: |
|

Genomic Map