Assay Details
Target Gene Details
Entrez Gene ID: | 66996 |
Gene Name: | carcinoembryonic antigen-related cell adhesion molecule 11 |
Gene Aliases: |
1600015D01Rik |
Location: |
Chr.7:17972167-17978556 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ceacam11 | NM_023289.2 | 3 | 726 | NP_075778.2 |
| AF221099.1 | 3 | 769 | AAG31946.1 | |
| AK004142.1 | 3 | 727 | BAB23191.1 | |
| AK005432.1 | 3 | 727 | BAB24026.1 | |
| AK005463.1 | 3 | 727 | BAB24054.1 | |
| AK145847.1 | 3 | 727 | BAE26694.1 | |
| BC099452.1 | 3 | 714 | AAH99452.1 |
More Information
Set Membership: |
|

Genomic Map