Assay Details
Target Gene Details
Entrez Gene ID: | 446099 |
Gene Name: | NLR family, pyrin domain containing 4E |
Gene Aliases: |
4930406H16Rik, Nalp-epsilon, Nalp4e, Nlrp4 |
Location: |
Chr.7:23301192-23400240 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nlrp4e | NM_001004194.2 | NP_001004194.2 | ||
AK019568.1 | BAC25598.1 | |||
AY596197.1 | AAU06318.1 | |||
BC139319.1 | ||||
BC139320.1 |
