Assay Details
Target Gene Details
Entrez Gene ID: | 236727 |
Gene Name: | solute carrier family 9 (sodium/hydrogen exchanger), member 7 |
Gene Aliases: |
A530087D17Rik, NHE7 |
Location: |
Chr.X:20098851-20291970 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc9a7 | NM_177353.3 | 1 | 82 | NP_796327.1 |
| AK041169.1 | 1 | 82 | BAC30848.1 | |
| AK043187.1 | 1 | 104 | ||
| AK078577.1 | 1 | 127 | ||
| BC058750.1 | 1 | 88 | AAH58750.1 |
More Information
Set Membership: |
|

Genomic Map