Assay Details
Target Gene Details
Entrez Gene ID: | 66768 |
Gene Name: | PARK2 co-regulated-like |
Gene Aliases: |
4933428G09Rik |
Location: |
Chr.5:48372392-48388756 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pacrgl | NM_025755.3 | NP_080031.1 | ||
| AK014276.1 | ||||
| AK016966.1 | BAB30527.1 | |||
| AK018101.1 | ||||
| AK032498.1 | BAC27898.1 | |||
| BC018481.1 | AAH18481.1 |

Genomic Map