Assay Details
Target Gene Details
Entrez Gene ID: | 73314 |
Gene Name: | leucine rich repeat containing 69 |
Gene Aliases: |
1700034K16Rik |
Location: |
Chr.4:14623622-14801320 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrrc69 | NM_028499.2 | NP_082775.1 | ||
| AK006601.1 | BAB24669.1 | |||
| AK161371.1 | BAE36353.1 |
More Information
Set Membership: |
|

Genomic Map