Assay Details
Target Gene Details
Entrez Gene ID: | 595136 |
Gene Name: | NADH dehydrogenase (ubiquinone) Fe-S protein 5 |
Gene Aliases: |
AI256693 |
Location: |
Chr.4:123712710-123718186 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufs5 | NM_001030274.1 | NP_001025445.1 | ||
| AK148505.1 | BAE28590.1 | |||
| AK152845.1 | BAE31538.1 | |||
| BC081434.1 | AAH81434.1 | |||
| BU534760.1 |
More Information
Set Membership: |
|

Genomic Map