Assay Details
Target Gene Details
Entrez Gene ID: | 195236 |
Gene Name: | POM121 membrane glycoprotein-like 2 (rat) |
Gene Aliases: |
Gm24 |
Location: |
Chr.13:21981181-21988734 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pom121l2 | NM_001162929.1 | 2 | 1735 | NP_001156401.1 |
| AK133022.1 | 2 | 1737 | BAE21474.1 |
More Information
Set Membership: |
|

Genomic Map